Learn More
Medchemexpress LLC Dazostinag disodium (TAK-676) | 2553413-93-5 | 98.65% | 754.48 | 100 UG
SDP
Catalog No. 5000361336
Encompass_Preferred
Dazostinag disodium (TAK-676) is a STING agonist that activates the STING signaling pathway and type I interferons. It also modulates the immune system, leading to complete regressions and durable memory T-cell immunity, promoting durable IFN-dependent antitumor immunity.
  • Activates the STING-TBK1-IRF3 pathway.
  • Modulates immune cells, including dendritic, natural killer, and T cells (CD8+, CD4+).
  • Well tolerated with dose-proportional pharmacokinetics.
  • Demonstrates anti-tumor function in syngeneic tumor models.
  • Induces cytokine responses and increases activation and proliferation of immune cells.
Catalog No. 50-003-61336 Supplier Medchemexpress LLC Supplier No. HY148029100UG
Dazostinag disodium (TAK-676) is a STING agonist that activates the STING signaling pathway and type I interferons. It also modulates the immune system, leading to complete regressions and durable memory T-cell immunity, promoting durable IFN-dependent antitumor immunity.
  • Activates the STING-TBK1-IRF3 pathway.
  • Modulates immune cells, including dendritic, natural killer, and T cells (CD8+, CD4+).
  • Well tolerated with dose-proportional pharmacokinetics.
  • Demonstrates anti-tumor function in syngeneic tumor models.
  • Induces cytokine responses and increases activation and proliferation of immune cells.