Learn More
Medchemexpress LLC HY-106067A 10mg Medchemexpress, Dazoxiben CAS:74226-22-5 Purity:>98%
SDP
Catalog No. 501968313
Encompass_Preferred
Medchemexpress, HY-106067A 10mg Dazoxiben CAS:74226-22-5 Dazoxiben is a potent and orally active thromboxane (TX) synthase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8313 Supplier Medchemexpress LLC Supplier No. HY106067A10MG
Medchemexpress, HY-106067A 10mg Dazoxiben CAS:74226-22-5 Dazoxiben is a potent and orally active thromboxane (TX) synthase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.