Learn More
Medchemexpress LLC Dazoxiben (hydrochloride) | 74226-22-5 | MFCD00866791 | 99.9% | 268.70 | C12H13ClN2O3 | 10 MM 1 ML
SDP
Catalog No. 5000242714
Encompass_Preferred
Dazoxiben (hydrochloride) is the hydrochloride salt of dazoxiben, a potent and orally active thromboxane (TX) synthase inhibitor used in research to probe thromboxane-mediated pathways. Supplied as a concentrated solution for laboratory use, it includes accompanying safety and handling documentation.
  • Potent thromboxane synthase inhibitor for research applications.
  • Provided as a 10 mM solution in DMSO, ready for dilution.
  • High purity (99.9%) suitable for biochemical assays.
  • Storage guidance for sealed samples: -80°C (6 months) or -20°C (1 month) in solvent.
  • SDS and data sheet available for handling and safety information.
Catalog No. 50-002-42714 Supplier Medchemexpress LLC Supplier No. HY106067A10MM 1ML
Dazoxiben (hydrochloride) is the hydrochloride salt of dazoxiben, a potent and orally active thromboxane (TX) synthase inhibitor used in research to probe thromboxane-mediated pathways. Supplied as a concentrated solution for laboratory use, it includes accompanying safety and handling documentation.
  • Potent thromboxane synthase inhibitor for research applications.
  • Provided as a 10 mM solution in DMSO, ready for dilution.
  • High purity (99.9%) suitable for biochemical assays.
  • Storage guidance for sealed samples: -80°C (6 months) or -20°C (1 month) in solvent.
  • SDS and data sheet available for handling and safety information.