Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dazzling Division
SCI_ED

Catalog No. S79230
Encompass
Encompass_Preferred
Change view
Click to view available options
Grade:
4 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S79230 4 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S79230 Supplier JOHN WILEY & SONS Supplier No. 0471369837
Add to Cart
Add to Cart

Get your students undivided attention!

To make division fun, Dazzling Division provides dozens of games and activities that use objects found in any household, such as playing cards, dice, coins, paper, and scissors. Lynette Long, soft cover, 128 pages. 2000.

Specifications

Grade 4 to 8
Format Book
Product Type Dazzling Division
For Use With (Application) Games and activities to make division fun such as playing cards, dice, coins, paper and scissors
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.