Learn More
Medchemexpress LLC DB-0646 | 2769753-59-3 | 98.0% | C52H51F3N14O7 | 10MG
SDP
Catalog No. 5000261753
Encompass_Preferred
DB-0646 is a proteolysis-targeting chimera (PROTAC) developed as a multi-kinase degrader for laboratory research. It induces ubiquitination and proteasomal degradation of target kinases and is supplied as a solid for biochemical and cellular studies. Intended for research use only; not for human or veterinary applications.
  • Multi-kinase degrader mechanism suitable for kinase-targeted studies.
  • High purity (98.0%) appropriate for in vitro assays.
  • Molecular weight 1041.05 with formula C52H51F3N14O7.
  • Available in small pack sizes for screening and optimization.
  • Stable under recommended storage conditions; follow supplier guidance.
This item is not returnable. View return policy
Catalog No. 50-002-61753 Supplier Medchemexpress LLC Supplier No. HY13734310MG
DB-0646 is a proteolysis-targeting chimera (PROTAC) developed as a multi-kinase degrader for laboratory research. It induces ubiquitination and proteasomal degradation of target kinases and is supplied as a solid for biochemical and cellular studies. Intended for research use only; not for human or veterinary applications.
  • Multi-kinase degrader mechanism suitable for kinase-targeted studies.
  • High purity (98.0%) appropriate for in vitro assays.
  • Molecular weight 1041.05 with formula C52H51F3N14O7.
  • Available in small pack sizes for screening and optimization.
  • Stable under recommended storage conditions; follow supplier guidance.