Learn More
Neta Scientific DB-1 15M 0.25MM 0.25UM LTM
SDP
Catalog No. 50927082
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-082 Supplier Neta Scientific Supplier No. 1221012LTM
This item is manufactured by Agilent