Learn More
Neta Scientific DB-1 15M 0.53MM 1.50UM LTM
SDP
Catalog No. 50927437
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-437 Supplier Neta Scientific Supplier No. 1251012LTM
This item is manufactured by Agilent