Learn More
Neta Scientific DB-1 30M 0.25MM 1.0UM LTM
SDP
Catalog No. 50927693
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-693 Supplier Neta Scientific Supplier No. 1221033LTM
This item is manufactured by Agilent