Learn More
Agilent Technologies J&W DB-1 GC Column, 30 m, 0.53 mm, 5.00 µm, 7 inch cage
Catalog No. NC2949394
Encompass
This is the most common GC column format compatible with Agilent 5890, 6890, 7820, 7890, 8860, and 8890 series GC systems and all non-Agilent GC systems with a similarly sized oven.
Catalog No. NC2949394 Supplier Agilent Technologies Supplier No. 1251035
This is the most common GC column format compatible with Agilent 5890, 6890, 7820, 7890, 8860, and 8890 series GC systems and all non-Agilent GC systems with a similarly sized oven.