Learn More
Neta Scientific DB-17 15M 0.32MM 0.50U
SDP
Catalog No. 50928604
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-604 Supplier Neta Scientific Supplier No. 1231713
This item is manufactured by Agilent