Learn More
Neta Scientific DB-17 30M 0.25MM 0.25UM LTM
SDP
Catalog No. 50927091
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-091 Supplier Neta Scientific Supplier No. 1221732LTM
This item is manufactured by Agilent