Learn More
Neta Scientific DB-17MS 30M 0.32MM 0.25U
SDP
Catalog No. 50928256
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-256 Supplier Neta Scientific Supplier No. 1234732
This item is manufactured by Agilent