Learn More
Neta Scientific DB-1HT 30M 0.53MM 0.17UM
SDP
Catalog No. 50928271
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-271 Supplier Neta Scientific Supplier No. 1251131
This item is manufactured by Agilent