Learn More
Neta Scientific DB-5 15M 0.32MM 0.10UM LTM
SDP
Catalog No. 50927404
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-404 Supplier Neta Scientific Supplier No. 1235011LTM
This item is manufactured by Agilent