Learn More
Neta Scientific DB-5 20M 0.18MM 0.40U
SDP
Catalog No. 50928670
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-670 Supplier Neta Scientific Supplier No. 1215023
This item is manufactured by Agilent