Learn More
Neta Scientific DB-5 25M 0.20MM 0.33U
SDP
Catalog No. 50928789
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-789 Supplier Neta Scientific Supplier No. 1285022
This item is manufactured by Agilent