Learn More
Neta Scientific DB-5 25M .32MM-1.05U
SDP
Catalog No. 50928921
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-921 Supplier Neta Scientific Supplier No. 123502F
This item is manufactured by Agilent