Learn More
Neta Scientific DB-5 25M .32MM-.25U
SDP
Catalog No. 50928939
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-939 Supplier Neta Scientific Supplier No. 1235022
This item is manufactured by Agilent