Learn More
Neta Scientific DB-5 50M 0.32MM 1.00U
SDP
Catalog No. 50928723
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-723 Supplier Neta Scientific Supplier No. 1235053
This item is manufactured by Agilent