Learn More
Neta Scientific DB-5 60M 0.25MM 0.25U
SDP
Catalog No. 50928689
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-689 Supplier Neta Scientific Supplier No. 1225062
This item is manufactured by Agilent