Learn More
Agilent Technologies DB-5 CAP COL 30MX0.25MMX0.25
Catalog No. NC1651186
Encompass
DB-5 Capillary column, 30 meters x 0.25 mm ID x 0.25 micron film
Catalog No. NC1651186 Supplier Agilent Technologies Supplier No. 1225032
DB-5 Capillary column, 30 meters x 0.25 mm ID x 0.25 micron film