Learn More
Neta Scientific DB-5MS 15M 0.25MM 0.10UM LTM
SDP
Catalog No. 50927724
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-724 Supplier Neta Scientific Supplier No. 1225511LTM
This item is manufactured by Agilent