Learn More
Neta Scientific DB-5MS 15M 0.53MM 1.50U
SDP
Catalog No. 50928492
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-492 Supplier Neta Scientific Supplier No. 1255512
This item is manufactured by Agilent