Learn More
Agilent Technologies DB-5Q Capillary Gas Chromatography Column, Fused Silica, 20 m, 0.18 mm ID, 0.18 μm Film Thickness
Catalog No. NC3982157
Encompass
The DB-5Q Ultra Low-Bleed GC/MS column is a capillary gas chromatography column designed for sensitive analysis. Constructed from high-temperature fused silica, this column offers a stationary phase with low polarity and is designated as G36 per USP standards.
  • Length of 20 m
  • Inner diameter of 0.18 mm
  • Film thickness of 0.18 μm
  • Temperature range of -60 °C to 350 °C
  • Designed for ultra low-bleed performance
  • Compatible with 7-inch cage oven systems
Catalog No. NC3982157 Supplier Agilent Technologies Supplier No. 1215522Q
The DB-5Q Ultra Low-Bleed GC/MS column is a capillary gas chromatography column designed for sensitive analysis. Constructed from high-temperature fused silica, this column offers a stationary phase with low polarity and is designated as G36 per USP standards.
  • Length of 20 m
  • Inner diameter of 0.18 mm
  • Film thickness of 0.18 μm
  • Temperature range of -60 °C to 350 °C
  • Designed for ultra low-bleed performance
  • Compatible with 7-inch cage oven systems