Learn More
Neta Scientific DB-FFAP 30M 0.32MM 0.25UM LTM
SDP
Catalog No. 50927398
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-398 Supplier Neta Scientific Supplier No. 1233232LTM
This item is manufactured by Agilent