Learn More
Neta Scientific DB-PETRO 50M 0.20MM 0.50U
SDP
Catalog No. 50928069
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-069 Supplier Neta Scientific Supplier No. 1281056
This item is manufactured by Agilent