Learn More
Neta Scientific DB-WAX 15M 0.32MM 0.25UM LTM
SDP
Catalog No. 50927786
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-786 Supplier Neta Scientific Supplier No. 1237012LTM
This item is manufactured by Agilent