Learn More
Neta Scientific DB-WAX 30M 0.53MM 0.25UM LTM
SDP
Catalog No. 50927840
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-840 Supplier Neta Scientific Supplier No. 1257031LTM
This item is manufactured by Agilent