Learn More
Medchemexpress LLC DB04760 | 544678-85-5 | 99.2% | 410.42 g·mol⁻¹ | C22H20F2N4O2 | 50MG
SDP
Catalog No. 5000235070
Encompass_Preferred
DB04760 is a potent, highly selective, non-zinc-chelating MMP-13 inhibitor (IC50 = 8 nM) used as a research reagent in preclinical studies. It has been reported to reduce paclitaxel-induced neurotoxicity and to exhibit anticancer activity. The compound has formula C22H20F2N4O2, molecular weight 410.42 g·mol⁻¹, and is supplied in solid and solution forms with high purity suitable for biological assays.
  • Potent MMP-13 inhibition with IC50 = 8 nM.
  • Non-zinc-chelating mechanism for selective target studies.
  • Shown to reduce paclitaxel-induced neurotoxicity in models.
  • Reported anticancer activity in preclinical research.
  • Available as solid and as 10 mM solution in DMSO for assay flexibility.
  • High purity (~99%) appropriate for biological testing.
Catalog No. 50-002-35070 Supplier Medchemexpress LLC Supplier No. HY12516650MG
DB04760 is a potent, highly selective, non-zinc-chelating MMP-13 inhibitor (IC50 = 8 nM) used as a research reagent in preclinical studies. It has been reported to reduce paclitaxel-induced neurotoxicity and to exhibit anticancer activity. The compound has formula C22H20F2N4O2, molecular weight 410.42 g·mol⁻¹, and is supplied in solid and solution forms with high purity suitable for biological assays.
  • Potent MMP-13 inhibition with IC50 = 8 nM.
  • Non-zinc-chelating mechanism for selective target studies.
  • Shown to reduce paclitaxel-induced neurotoxicity in models.
  • Reported anticancer activity in preclinical research.
  • Available as solid and as 10 mM solution in DMSO for assay flexibility.
  • High purity (~99%) appropriate for biological testing.