Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DB04760 (compound 4) | 544678-85-5 | MFCD09878513 | 99.2% | 410.42 g/mol | C22H20F2N4O2 | 10MG
SDP

Catalog No. 5000237321
Encompass_Preferred

DB04760 is a small-molecule MMP-13 inhibitor (compound 4) reported as a potent, highly selective, non-zinc-chelating inhibitor with an IC50 of 8 nM. It has been used in studies showing reduction of paclitaxel-induced neurotoxicity and anticancer activity, and is supplied for research use with defined purity and solubility characteristics.

  • Potent MMP-13 inhibitor with IC50 = 8 nM.
  • Highly selective non-zinc-chelating mechanism.
  • Molecular formula C22H20F2N4O2, molecular weight 410.42 g/mol.
  • CAS number 544678-85-5.
  • Purity 99.2%.
  • Soluble in DMSO at ≥100 mg/mL; in vivo formulation: 10% DMSO then 90% corn oil.
  • Reported to reduce paclitaxel neurotoxicity and exhibit anticancer activity.
  • Available in multiple small-scale pack sizes for research use.

Catalog No. 50-002-37321 Supplier Medchemexpress LLC Supplier No. HY12516610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DB04760 is a small-molecule MMP-13 inhibitor (compound 4) reported as a potent, highly selective, non-zinc-chelating inhibitor with an IC50 of 8 nM. It has been used in studies showing reduction of paclitaxel-induced neurotoxicity and anticancer activity, and is supplied for research use with defined purity and solubility characteristics.

  • Potent MMP-13 inhibitor with IC50 = 8 nM.
  • Highly selective non-zinc-chelating mechanism.
  • Molecular formula C22H20F2N4O2, molecular weight 410.42 g/mol.
  • CAS number 544678-85-5.
  • Purity 99.2%.
  • Soluble in DMSO at ≥100 mg/mL; in vivo formulation: 10% DMSO then 90% corn oil.
  • Reported to reduce paclitaxel neurotoxicity and exhibit anticancer activity.
  • Available in multiple small-scale pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.