Learn More
Medchemexpress LLC HY-15737 5mg Medchemexpress, DB07268 CAS:929007-72-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY157375MG
Encompass_Preferred
Medchemexpress, HY-15737 5mg DB07268 CAS:929007-72-7 DB07268 is a potent and selective JNK1 inhibitor with an IC50 value of 9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1785