Learn More
TARGETMOL CHEMICALS INC DB1976 5MG
SDP
Catalog No. 502581611
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DB1976 is a selenophene analog of DB270 and a potent and cell-permeable fully efficacious transcription factor PU.1 inhibitor. DB1976 potently inhibits PU.1 binding (IC50 of 10 nM) and strongly inhibits the PU.1/DNA complex (with high DB1976-(lambda)B affinity KD of 12 nM) in vitro. DB1976 has Apoptosis-inducing effect. purity: 98%
Catalog No. 50-258-1611 Supplier TARGETMOL CHEMICALS INC Supplier No. T109645MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DB1976 is a selenophene analog of DB270 and a potent and cell-permeable fully efficacious transcription factor PU.1 inhibitor. DB1976 potently inhibits PU.1 binding (IC50 of 10 nM) and strongly inhibits the PU.1/DNA complex (with high DB1976-(lambda)B affinity KD of 12 nM) in vitro. DB1976 has Apoptosis-inducing effect. purity: 98%