Learn More
Medchemexpress LLC DB2115 tertahydrochloride | 1366126-19-3 | 99.1% | 704.48 | C32H34Cl4N8O2 | 5MG
SDP
Catalog No. 5000235436
Encompass_Preferred
DB2115 tertahydrochloride is a small-molecule inhibitor of the transcription factor PU.1 supplied as a tertahydrochloride salt for research use. It suppresses PU.1 binding to DNA (IC50: 2.3 nM), exhibits antiproliferative and pro-apoptotic activity in tumor cell models, and is provided as a solid powder suitable for biochemical and cellular studies.
  • Highly potent PU.1 inhibition (IC50: 2.3 nM).
  • Demonstrates antiproliferative and pro-apoptotic effects in tumor cells.
  • High purity for research applications.
  • Available in multiple small-scale pack sizes for experimental flexibility.
  • Stable when stored sealed at recommended temperatures.
Catalog No. 50-002-35436 Supplier Medchemexpress LLC Supplier No. HY124676A5MG
DB2115 tertahydrochloride is a small-molecule inhibitor of the transcription factor PU.1 supplied as a tertahydrochloride salt for research use. It suppresses PU.1 binding to DNA (IC50: 2.3 nM), exhibits antiproliferative and pro-apoptotic activity in tumor cell models, and is provided as a solid powder suitable for biochemical and cellular studies.
  • Highly potent PU.1 inhibition (IC50: 2.3 nM).
  • Demonstrates antiproliferative and pro-apoptotic effects in tumor cells.
  • High purity for research applications.
  • Available in multiple small-scale pack sizes for experimental flexibility.
  • Stable when stored sealed at recommended temperatures.