Learn More
Medchemexpress LLC DB2313 | 2170606-74-1 | MFCD34179477 | 99.5% | 708.83 | C42H41FN8O2 | 250MG
SDP
Catalog No. 5000238084
Encompass_Preferred
DB2313 is a small-molecule inhibitor of the PU.1 transcription factor used for preclinical research. It disrupts PU.1 interaction with target gene promoters, inhibits PU.1-dependent reporter gene transactivation (IC50 ≈ 5 μM), and induces apoptosis in acute myeloid leukemia cell models. The compound is supplied as a high-purity solid for in vitro studies; chemical formula C42H41FN8O2, molecular weight 708.83.
Catalog No. 50-002-38084 Supplier Medchemexpress LLC Supplier No. HY124629250MG
DB2313 is a small-molecule inhibitor of the PU.1 transcription factor used for preclinical research. It disrupts PU.1 interaction with target gene promoters, inhibits PU.1-dependent reporter gene transactivation (IC50 ≈ 5 μM), and induces apoptosis in acute myeloid leukemia cell models. The compound is supplied as a high-purity solid for in vitro studies; chemical formula C42H41FN8O2, molecular weight 708.83.