Learn More
Medchemexpress LLC HY-124629 5mg , DB2313 CAS:2170606-74-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246295MG
Encompass_Preferred
HY-124629 5mg DB2313 CAS: 2170606-74-1 DB2313 is a potent transcription factor PU.1 inhibitor with an apoptosis of 14 nM. DB2313 disrupts the interaction of PU.1 with target gene promoters. DB2313 induces apoptosis of acute myeloid leukemia (AML) cells, and has anticancer effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9176