Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex DB25 Male Connector; for Creating Your Own Cable
Catalog No. NC0097310
Encompass
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. The maximum cable length is 1,000 feet or 300 meters.
Catalog No. NC0097310 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0752394
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. The maximum cable length is 1,000 feet or 300 meters.