Learn More
Cole-Parmer Dri-Block Analog Heater, triple insert, 100°C; 115 V
Supplier:  Cole-Parmer 9996882
Encompass
Techne Dri-Block Analog Block Heater Triple Insert 100°C 115 VAC, Precision-engineered heaters with excellent temperature accuracyVersatile range of interchangeable heating blocks work with many size sample tubes and platesTake up less storage space—lightweight and flat-top surface allow units to be stacked when not in use115 VAC operation
Catalog No. NC1129643