Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience DB50 Beakers
SDP

Catalog No. 16948949 Shop All Interscience Products
Change view
Click to view available options
Graduated:
No
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Graduated
16-948-949 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-948-949 Supplier Interscience Supplier No. 415100
Add to Cart
Add to Cart

DB50 Beakers for EasySpiral Dilute

  • Beakers for use with EasySpiral Dilute
  • Volume: 5 mL
  • Quantity: 1,000 beakers

Specifications

Color Clear
Capacity (Metric) 5 mL
For Use With (Equipment) EasySpiral Dilute
Quantity 1000/Cs.
Type EasySpiral Beakers
Graduated No
Scale Not Graduated
Style Nesting
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.