Learn More
Medchemexpress LLC Db772 (hydrate) | 99.9% | 502.54 g·mol⁻¹ | C20H19ClN4O2 | 5 MG
SDP
Catalog No. 5000263124
Encompass_Preferred
DB772 hydrate is a research-grade small molecule described as an inhibitor of bovine viral diarrhoea virus (BVDV) that also exhibits anti-prion activity. Supplied as a hydrated solid for laboratory use, it is intended for in vitro research only and is not for human or diagnostic use.
  • Inhibits bovine viral diarrhoea virus (BVDV) in vitro.
  • Demonstrates anti-prion activity in reported studies.
  • Supplied as a hydrated solid suitable for laboratory assays.
  • High purity suitable for research applications.
  • Available in multiple milligram pack sizes for flexibility.
Catalog No. 50-002-63124 Supplier Medchemexpress LLC Supplier No. HY114621A5MG
DB772 hydrate is a research-grade small molecule described as an inhibitor of bovine viral diarrhoea virus (BVDV) that also exhibits anti-prion activity. Supplied as a hydrated solid for laboratory use, it is intended for in vitro research only and is not for human or diagnostic use.
  • Inhibits bovine viral diarrhoea virus (BVDV) in vitro.
  • Demonstrates anti-prion activity in reported studies.
  • Supplied as a hydrated solid suitable for laboratory assays.
  • High purity suitable for research applications.
  • Available in multiple milligram pack sizes for flexibility.