Learn More
Medchemexpress LLC HY-121805 25mg Medchemexpress, DBCO-​C6-​acid CAS:1425485-72-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12180525MG
Encompass_Preferred
Medchemexpress, HY-121805 25mg DBCO-​C6-​acid CAS:1425485-72-8 DBCO-C6-acid is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). DBCO-C6-acid can be used in the synthesis of carmaphycin analogues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0976