Learn More
Medchemexpress LLC HY-42972 10mg Medchemexpress, DBCO-acid CAS:1353016-70-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY4297210MG
Encompass_Preferred
Medchemexpress, HY-42972 10mg DBCO-acid CAS:1353016-70-2 DBCO-acid is a cleavable ADC linker used in the synthesis of ADC linker DBCO-NHS ester (HY-115524 and HY-115545), and drug-linker conjugates DBCO-PEG-MMAE (HY-111012 and HY-126690). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0056