Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-acid | 1353016-70-2 | 99.7% | 305.33 g/mol | C19H15NO3 | 250 MG
SDP

Catalog No. 5000204078
Encompass_Preferred

DBCO-acid is a strain-promoted alkyne reagent used in bioorthogonal click chemistry (SPAAC) to conjugate azide-containing molecules. It is commonly applied in the synthesis of antibody-drug conjugate linkers and agent-linker conjugates.

  • Strain-promoted alkyne reagent for SPAAC conjugation to azide-containing molecules.
  • Commonly used to synthesize ADC linkers and agent-linker conjugates.
  • Chemical formula C19H15NO3; molecular weight 305.33 g/mol.
  • Purity 99.7%; CAS number 1353016-70-2.
  • Soluble in DMSO (~100 mg/mL); hygroscopic-use freshly opened DMSO.
  • Store at 4°C under inert gas; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-04078 Supplier Medchemexpress LLC Supplier No. HY42972250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-acid is a strain-promoted alkyne reagent used in bioorthogonal click chemistry (SPAAC) to conjugate azide-containing molecules. It is commonly applied in the synthesis of antibody-drug conjugate linkers and agent-linker conjugates.

  • Strain-promoted alkyne reagent for SPAAC conjugation to azide-containing molecules.
  • Commonly used to synthesize ADC linkers and agent-linker conjugates.
  • Chemical formula C19H15NO3; molecular weight 305.33 g/mol.
  • Purity 99.7%; CAS number 1353016-70-2.
  • Soluble in DMSO (~100 mg/mL); hygroscopic-use freshly opened DMSO.
  • Store at 4°C under inert gas; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.