Learn More
Medchemexpress LLC DBCO-amine | 1255942-06-3 | 99.1% | 276.34 g·mol-1 | C18H16N2O | 25 MG
SDP
Catalog No. 5000202364
Encompass_Preferred
DBCO-amine is a dibenzocyclooctyne-containing amine reagent used for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry. It is used as a small linker for bioconjugation and the synthesis of antibody-drug conjugates, and is typically handled and stored under anhydrous conditions and in organic solvent solutions for formulation.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) bioconjugation.
  • High purity suitable for synthetic and bioconjugation applications.
  • Minimal spacer length reduces steric hindrance in conjugates.
  • Compatible with common coupling chemistries for linker installation.
  • Soluble in DMSO for straightforward formulation and handling.
Catalog No. 50-002-02364 Supplier Medchemexpress LLC Supplier No. HYW00042325MG
DBCO-amine is a dibenzocyclooctyne-containing amine reagent used for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry. It is used as a small linker for bioconjugation and the synthesis of antibody-drug conjugates, and is typically handled and stored under anhydrous conditions and in organic solvent solutions for formulation.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) bioconjugation.
  • High purity suitable for synthetic and bioconjugation applications.
  • Minimal spacer length reduces steric hindrance in conjugates.
  • Compatible with common coupling chemistries for linker installation.
  • Soluble in DMSO for straightforward formulation and handling.