Learn More
TARGETMOL CHEMICALS INC DBCO-amine 25MG
SDP
Catalog No. 502880038
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DBCO-amine is a cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs)[1]. Purity 97.73%
Catalog No. 50-288-0038 Supplier TARGETMOL CHEMICALS INC Supplier No. T1505825MG
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DBCO-amine is a cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs)[1]. Purity 97.73%