Learn More
Medchemexpress LLC DBCO-amine (GMP) | 1255942-06-3 | 276.33 | 1 G
SDP
Catalog No. 5000447110
Encompass_Preferred
DBCO-amine (GMP) is a GMP grade click chemistry reagent that contains a DBCO group. It undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups, making it suitable for the synthesis of antibody-drug conjugates (ADCs).
  • Click chemistry reagent
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Suitable for synthesis of antibody-drug conjugates (ADCs)
  • GMP grade
Catalog No. 50-004-47110 Supplier Medchemexpress LLC Supplier No. HYW000423G1G
DBCO-amine (GMP) is a GMP grade click chemistry reagent that contains a DBCO group. It undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups, making it suitable for the synthesis of antibody-drug conjugates (ADCs).
  • Click chemistry reagent
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Suitable for synthesis of antibody-drug conjugates (ADCs)
  • GMP grade