Learn More
Medchemexpress LLC DBCO-amine (GMP) | 1255942-06-3 | 276.33 | 100 MG
SDP
Catalog No. 5000446624
Encompass_Preferred
DBCO-amine (GMP) is a GMP grade DBCO-amine. It is a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. It is used in the synthesis of antibody-drug conjugates (ADCs).
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • For research use only
Catalog No. 50-004-46624 Supplier Medchemexpress LLC Supplier No. HYW000423G100MG
DBCO-amine (GMP) is a GMP grade DBCO-amine. It is a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. It is used in the synthesis of antibody-drug conjugates (ADCs).
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • For research use only