Learn More
Medchemexpress LLC Dbco-amine (Gmp) | 1255942-06-3 | 276.33 | 500 MG
SDP
Catalog No. 5000446623
Encompass_Preferred
DBCO-amine (GMP) is a GMP grade click chemistry reagent that contains a DBCO group. This group enables strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules that possess azide groups. It is utilized in the synthesis of antibody-drug conjugates (ADCs).
  • Gmp grade
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC reactions
  • Used in the synthesis of antibody-drug conjugates (ADCs)
Catalog No. 50-004-46623 Supplier Medchemexpress LLC Supplier No. HYW000423G500MG
DBCO-amine (GMP) is a GMP grade click chemistry reagent that contains a DBCO group. This group enables strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules that possess azide groups. It is utilized in the synthesis of antibody-drug conjugates (ADCs).
  • Gmp grade
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC reactions
  • Used in the synthesis of antibody-drug conjugates (ADCs)