Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-amine (GMP) | 1255942-06-3 | MFCD22380759 | 276.33 | 200 MG
SDP

Catalog No. 5000447539
Encompass_Preferred

DBCO-amine (GMP) is a high-grade click chemistry reagent featuring a DBCO group, enabling it to participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with azide-containing molecules. This reagent is particularly utilized in the synthesis of antibody-drug conjugates (ADCs). It is a white to off-white solid.

  • It is a click chemistry reagent.
  • Contains a DBCO group.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • Used in the synthesis of antibody-drug conjugates.
  • Appearance: solid.
  • Color: white to off-white.

Catalog No. 50-004-47539 Supplier Medchemexpress LLC Supplier No. HYW000423G200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-amine (GMP) is a high-grade click chemistry reagent featuring a DBCO group, enabling it to participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with azide-containing molecules. This reagent is particularly utilized in the synthesis of antibody-drug conjugates (ADCs). It is a white to off-white solid.

  • It is a click chemistry reagent.
  • Contains a DBCO group.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • Used in the synthesis of antibody-drug conjugates.
  • Appearance: solid.
  • Color: white to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.