Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-biotin | 1418217-95-4 | MFCD22380751 | 98.5% | 502.63 | C28H30N4O3S | 50MG
SDP

Catalog No. 5000238822
Encompass_Preferred

DBCO-Biotin is an alkyl chain-based PROTAC linker and click-chemistry reagent that contains a dibenzocyclooctyne (DBCO) moiety conjugated to biotin, enabling copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. Supplied as a white to off-white solid, it is used for biotinylation, conjugation, and PROTAC synthesis in chemical biology applications.

  • High purity of 98.5%.
  • Molecular weight 502.63 g/mol.
  • Molecular formula C28H30N4O3S.
  • Soluble in DMSO at 31.25 mg/mL (62.17 mM).
  • Store protected from light under inert gas; solid recommended at -20°C.
  • Packaged as 50 mg vial.
  • Enables copper-free click conjugation via SPAAC.

Catalog No. 50-002-38822 Supplier Medchemexpress LLC Supplier No. HY12391650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-Biotin is an alkyl chain-based PROTAC linker and click-chemistry reagent that contains a dibenzocyclooctyne (DBCO) moiety conjugated to biotin, enabling copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. Supplied as a white to off-white solid, it is used for biotinylation, conjugation, and PROTAC synthesis in chemical biology applications.

  • High purity of 98.5%.
  • Molecular weight 502.63 g/mol.
  • Molecular formula C28H30N4O3S.
  • Soluble in DMSO at 31.25 mg/mL (62.17 mM).
  • Store protected from light under inert gas; solid recommended at -20°C.
  • Packaged as 50 mg vial.
  • Enables copper-free click conjugation via SPAAC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.