Learn More
Medchemexpress LLC DBCO-biotin | 1418217-95-4 | MFCD22380751 | 98.5% | 502.63 g·mol⁻1 | C28H30N4O3S | 10MG
SDP
Catalog No. 5000235793
Encompass_Preferred
DBCO-Biotin is a bifunctional click-chemistry reagent that links a dibenzocyclooctyne (DBCO) reactive moiety to a biotin affinity tag, enabling copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) for bioconjugation and probe development. Supplied as a high-purity solid for laboratory research, it is commonly used in bioconjugation, affinity enrichment, and linker synthesis applications.
  • Enables copper-free click chemistry via DBCO-mediated SPAAC.
  • Provides a biotin tag for affinity capture and detection.
  • High purity suitable for sensitive bioconjugation workflows.
  • Available in small mg-scale quantities for research use.
  • Stable when stored under recommended conditions; consult safety data for handling.
Catalog No. 50-002-35793 Supplier Medchemexpress LLC Supplier No. HY12391610MG
DBCO-Biotin is a bifunctional click-chemistry reagent that links a dibenzocyclooctyne (DBCO) reactive moiety to a biotin affinity tag, enabling copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) for bioconjugation and probe development. Supplied as a high-purity solid for laboratory research, it is commonly used in bioconjugation, affinity enrichment, and linker synthesis applications.
  • Enables copper-free click chemistry via DBCO-mediated SPAAC.
  • Provides a biotin tag for affinity capture and detection.
  • High purity suitable for sensitive bioconjugation workflows.
  • Available in small mg-scale quantities for research use.
  • Stable when stored under recommended conditions; consult safety data for handling.